Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URH1
Entrez Gene ID
818204
Gene Name
uridine-ribohydrolase 1
Gene Aliases
AT2G36310, F2H17.8, F2H17_8, NSH1, nucleoside hydrolase 1, uridine-ribohydrolase 1
Chromosome Location
Chr.2: 15224497 - 15227276 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129188.4 NP_565843.1 4 - 5 1103 65
GenBank mRNA AY054182.1 - 4 - 5 554 65
AY066040.1 - 4 - 5 460 65
AY086551.1 - 4 - 5 583 65

View Full Product Details