Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLS6
Entrez Gene ID
834416
Gene Name
flavonol synthase 6
Gene Aliases
AT5G43935, ATFLS6, flavonol synthase 6
Chromosome Location
Chr.5: 17682179 - 17683446 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148083.1 NP_680388.1 1 - 2 385 84

View Full Product Details