Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBD3L1
Entrez Gene ID
768027
Gene Name
methyl-CpG binding domain protein 3 like 1
Gene Aliases
-
Chromosome Location
Chr.17: 44256808 - 44259227 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077086.2 NP_001070554.1 1 - 2 81 145
GenBank mRNA BC112533.1 - 1 - 2 82 145

View Full Product Details