Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH3B1
Entrez Gene ID
511469
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
-
Chromosome Location
Chr.29: 45537445 - 45557502 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075518.1 NP_001068986.1 2 - 3 757 72
GenBank mRNA BC147958.1 - 3 - 4 303 72
BT025453.1 - 2 - 3 757 72
BT026328.1 - 3 - 4 557 72

View Full Product Details