Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REXO2
Entrez Gene ID
540139
Gene Name
RNA exonuclease 2
Gene Aliases
-
Chromosome Location
Chr.15: 24957716 - 24968465 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081735.2 NP_001075204.1 4 - 5 487 131
GenBank mRNA BC133575.1 - 4 - 5 488 131

View Full Product Details