Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC13
Entrez Gene ID
538716
Gene Name
homeobox C13
Gene Aliases
-
Chromosome Location
Chr.5: 26085810 - 26093573 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083490.1 NP_001076959.1 1 - 2 776 94
GenBank mRNA BC123829.1 - 1 - 2 776 94

View Full Product Details