Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF1AY
Entrez Gene ID
100271756
Gene Name
eukaryotic translation initiation factor 1A, Y-linked
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145757.1 NP_001139229.1 - 29 67

View Full Product Details