The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145757.1 | NP_001139229.1 | - | 275 | 139 |
GenBank mRNA | FJ627277.1 | - | - | 266 | 139 |