Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIN3A
Entrez Gene ID
100295209
Gene Name
glutamate ionotropic receptor NMDA type subunit 3A
Gene Aliases
-
Chromosome Location
Chr.8: 91311443 - 91511994 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191535.1 NP_001178464.1 7 - 8 2927 95

View Full Product Details