Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRB1
Entrez Gene ID
524070
Gene Name
adhesion G protein-coupled receptor B1
Gene Aliases
BAI1
Chromosome Location
Chr.14: 1797345 - 1874927 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191541.1 NP_001178470.1 22 - 23 3300 89

View Full Product Details