Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EAF1
Entrez Gene ID
507577
Gene Name
ELL associated factor 1
Gene Aliases
-
Chromosome Location
Chr.1: 152450841 - 152463273 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192153.3 NP_001179082.2 1 - 2 374 84
GenBank mRNA BC149556.1 - 1 - 2 258 84

View Full Product Details