Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSC
Entrez Gene ID
514862
Gene Name
goosecoid homeobox
Gene Aliases
goosecoid
Chromosome Location
Chr.21: 59628000 - 59629562 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192386.1 NP_001179315.1 2 - 3 620 64
GenBank mRNA JF919675.1 - 2 - 3 610 64

View Full Product Details