Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERCC4
Entrez Gene ID
516371
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.25: 12833389 - 12874857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205360.1 NP_001192289.1 7 - 8 1218 90

View Full Product Details