Product Details
- SNP ID
-
rs74345264
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:91369960 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCATGTTTAGCAAATTGTTCTT[C/T]AGGTAATGAAAAACAGTATTCTCAT
- Phenotype
-
MIM: 612464
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARRDC3
PubMed Links
Gene Details
- Gene
- ARRDC3
- Gene Name
- arrestin domain containing 3
There are no transcripts associated with this gene.
View Full Product Details