Product Details
- SNP ID
-
rs76946816
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:50009553 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAGTAACAACGAGAAAGTGACAC[A/G]CAATTTGGTACTTAATTATATGTAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB110
PubMed Links
Gene Details
- Gene
- DEFB110
- Gene Name
- defensin beta 110
View Full Product Details