Product Details

SNP ID
rs75008303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130345527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCTATTCCCAAACTATAAAGG[G/T]AACTGTGTGGGGCTTCAGCAGGACT
Phenotype
MIM: 609561
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPA5 PubMed Links

Gene Details

Gene
CPA5
Gene Name
carboxypeptidase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127441.1 737 Intron NP_001120913.1
NM_001127442.1 737 Intron NP_001120914.1
NM_001318223.1 737 Intron NP_001305152.1
NM_080385.4 737 Intron NP_525124.3
XM_005250710.1 737 Intron XP_005250767.1
XM_005250712.1 737 Intron XP_005250769.1
XM_011516698.1 737 UTR 5 XP_011515000.1
XM_011516701.1 737 UTR 5 XP_011515003.1
XM_011516702.1 737 UTR 5 XP_011515004.1
XM_011516703.1 737 UTR 5 XP_011515005.1
XM_011516704.1 737 UTR 5 XP_011515006.1
XM_011516705.1 737 UTR 5 XP_011515007.1
Gene
LOC101928632
Gene Name
uncharacterized LOC101928632
There are no transcripts associated with this gene.

View Full Product Details