Product Details

SNP ID
rs79059071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124868129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGTTTCCCACTATGGGAGTCTG[C/T]ATCCTTACCACCGAAGATAAGGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030978.2 Intron NP_112240.1
XM_011519067.1 Intron XP_011517369.1
Gene
RPL35
Gene Name
ribosomal protein L35
There are no transcripts associated with this gene.

View Full Product Details