Product Details

SNP ID
rs75762326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18414481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTTCCTGTTTGGGCCATAACGA[C/T]GCATACTAAAAGCAGCTTTAGCCAA
Phenotype
MIM: 150000 MIM: 150150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDHA PubMed Links

Gene Details

Gene
LDHA
Gene Name
lactate dehydrogenase A
There are no transcripts associated with this gene.

Gene
LDHC
Gene Name
lactate dehydrogenase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002301.4 Intron NP_002292.1
NM_017448.3 Intron NP_059144.1
XM_017017721.1 Intron XP_016873210.1
XM_017017722.1 Intron XP_016873211.1

View Full Product Details