Product Details

SNP ID
rs76039547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68069295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAGTATTCTTTGTGGCTATAG[A/G]TATTCTCTGCTAAATCTTGCAAACA
Phenotype
MIM: 603566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIAS1 PubMed Links

Gene Details

Gene
PIAS1
Gene Name
protein inhibitor of activated STAT 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320687.1 Intron NP_001307616.1
NM_016166.2 Intron NP_057250.1
XM_011522126.2 Intron XP_011520428.1
XM_011522127.2 Intron XP_011520429.1
XM_017022688.1 Intron XP_016878177.1
XM_017022689.1 Intron XP_016878178.1
XM_017022690.1 Intron XP_016878179.1
XM_017022691.1 Intron XP_016878180.1

View Full Product Details