Product Details

SNP ID
rs74567942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19257376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTGTCCCACGTGACCGCAAGG[A/G]ACTAGTGTTCAGTCTCCAGTGAGAT
Phenotype
MIM: 600826 MIM: 606563
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAPLN4 PubMed Links

Gene Details

Gene
HAPLN4
Gene Name
hyaluronan and proteoglycan link protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023002.2 1705 UTR 3 NP_075378.1
Gene
NCAN
Gene Name
neurocan
There are no transcripts associated with this gene.

Gene
TM6SF2
Gene Name
transmembrane 6 superfamily member 2
There are no transcripts associated with this gene.

View Full Product Details