Product Details

SNP ID
rs76878315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19636471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAAGCAGAGAATTAAATTCTAA[C/T]AGTGTATATTAATTTCCTTTTTTTT
Phenotype
MIM: 611626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPHOSPH8 PubMed Links

Gene Details

Gene
MPHOSPH8
Gene Name
M-phase phosphoprotein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017520.3 Intron NP_059990.2
XM_011535124.2 Intron XP_011533426.1

View Full Product Details