Product Details

SNP ID
rs79708098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43376257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCTTATCCTCAAGGAAGGACT[G/T]CTCTCCAAGGGCAGGCTGTTAGGCC
Phenotype
MIM: 616926 MIM: 601037
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FXYD4 PubMed Links

Gene Details

Gene
FXYD4
Gene Name
FXYD domain containing ion transport regulator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184963.1 693 UTR 3 NP_001171892.1
NM_173160.2 693 UTR 3 NP_775183.1
Gene
HNRNPF
Gene Name
heterogeneous nuclear ribonucleoprotein F
There are no transcripts associated with this gene.

View Full Product Details