Product Details
- SNP ID
-
rs78953026
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:247036819 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACAATTCTTTATTAGCAAAGAAA[A/G]CAGAAGAAAATCTTTACAACAAAAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF670
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs570580244] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF670
- Gene Name
- zinc finger protein 670
- Gene
- ZNF670-ZNF695
- Gene Name
- ZNF670-ZNF695 readthrough (NMD candidate)
There are no transcripts associated with this gene.
View Full Product Details