Product Details

SNP ID
rs115518996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:63727563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGCTATACCTCATTCCTAGAGG[A/G]CCTGATTTATTAAAGCCAGGATGGA
Phenotype
MIM: 612424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EYS PubMed Links

Gene Details

Gene
EYS
Gene Name
eyes shut homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142800.1 Intron NP_001136272.1
NM_001142801.1 Intron NP_001136273.1
NM_001292009.1 Intron NP_001278938.1
NM_198283.1 Intron NP_938024.1

View Full Product Details