Product Details

SNP ID
rs140057395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66003706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAATGGCTGAAAGACACTTGTGG[C/T]GCCAACGCCAAGCAGTCCCGGGACT
Phenotype
MIM: 603811 MIM: 601891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143985.1 705 Silent Mutation GGC,GGT G68G NP_001137457.1
NM_003860.3 705 Silent Mutation GGC,GGT G68G NP_003851.1
XM_017018514.1 705 Silent Mutation GGC,GGT G68G XP_016874003.1
XM_017018515.1 705 Intron XP_016874004.1
Gene
CST6
Gene Name
cystatin E/M
There are no transcripts associated with this gene.

Gene
EIF1AD
Gene Name
eukaryotic translation initiation factor 1A domain containing
There are no transcripts associated with this gene.

View Full Product Details