Product Details

SNP ID
rs151074449
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130003873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTTATTTTGAAGATTAAGTGAGC[C/T]CTTACTTTACCGAGTAAAGTGCTTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LINC00167 PubMed Links

Gene Details

Gene
LINC00167
Gene Name
long intergenic non-protein coding RNA 167
There are no transcripts associated with this gene.

Gene
PRDM10
Gene Name
PR domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020228.2 Intron NP_064613.2
NM_199437.1 Intron NP_955469.1
NM_199438.1 Intron NP_955470.1
NM_199439.1 Intron NP_955471.1
XM_011542912.2 Intron XP_011541214.1
XM_017018023.1 Intron XP_016873512.1

View Full Product Details