Product Details

SNP ID
rs142303164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52544563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTTTTCTTGGAGGGTGCACTC[A/G]GGCTGGAACCCTTCCCTAGGGTGTC
Phenotype
MIM: 608245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT71 PubMed Links

Gene Details

Gene
KRT71
Gene Name
keratin 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033448.2 1611 Missense Mutation CCG,CTG P514L NP_258259.1
XM_017018749.1 1611 Missense Mutation CCG,CTG P432L XP_016874238.1

View Full Product Details