Product Details

SNP ID
rs141027642
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102730637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGGAATTTTCTGATGAGTGAT[A/G]AGTTTTCATGGGGACTTTGCTTTTT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 20200 Missense Mutation TCA,TTA S6687L NP_001139669.1
XM_011521106.1 20200 Missense Mutation TCA,TTA S6647L XP_011519408.1

View Full Product Details