Product Details

SNP ID
rs144805209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102730719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGGAAAGACTACCACTTTGTC[C/T]ACCAGCATTTACGTTTTTACACACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 20118 Missense Mutation AGA,GGA R6660G NP_001139669.1
XM_011521106.1 20118 Missense Mutation AGA,GGA R6620G XP_011519408.1

View Full Product Details