Product Details

SNP ID
rs143388121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:29577412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATGGGTGATGTAGCGCTCCCC[A/G]ATTTTGCATTCCAGCTCTCGCAAAT
Phenotype
MIM: 605435
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKD1 PubMed Links

Gene Details

Gene
PRKD1
Gene Name
protein kinase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002742.2 5126 Silent Mutation ATC,ATT I855I NP_002733.2
XM_005267859.1 5126 Silent Mutation ATC,ATT I863I XP_005267916.1
XM_011536964.2 5126 Silent Mutation ATC,ATT I787I XP_011535266.1
XM_011536965.2 5126 Silent Mutation ATC,ATT I767I XP_011535267.1
XM_017021461.1 5126 Silent Mutation ATC,ATT I767I XP_016876950.1
XM_017021462.1 5126 Silent Mutation ATC,ATT I690I XP_016876951.1

View Full Product Details