Product Details
- SNP ID
-
rs148651064
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:59207056 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTCAAGCAACGCGCATCCCGCTCA[A/G]CGGCACCTGGCTCTTCACCCCCGTG
- Phenotype
-
MIM: 601479
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LDHAL6B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3809529] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LDHAL6B
- Gene Name
- lactate dehydrogenase A like 6B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033195.2 |
241 |
Missense Mutation |
AAC,AGC |
N39S |
NP_149972.1 |
- Gene
- MYO1E
- Gene Name
- myosin IE
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004998.3 |
241 |
Intron |
|
|
NP_004989.2 |
View Full Product Details