Product Details

SNP ID
rs140051221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41008929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCAGCAGGGTGGGGGGCAGGTG[C/T]CACAGCAGATGGGCTGAAGGAGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP3-1 PubMed Links

Gene Details

Gene
KRTAP3-1
Gene Name
keratin associated protein 3-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031958.1 186 Missense Mutation GAC,GGC D49G NP_114164.1
Gene
KRTAP3-2
Gene Name
keratin associated protein 3-2
There are no transcripts associated with this gene.

View Full Product Details