Product Details
- SNP ID
-
rs139243712
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35939456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCCTCCCGTTGCTCCTGTGCCTG[C/G]TGCCGCTGGCCCCTGCCTCATCCCC
- Phenotype
-
MIM: 612809
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105372383
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2242590] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105372383
- Gene Name
- uncharacterized LOC105372383
There are no transcripts associated with this gene.
- Gene
- LRFN3
- Gene Name
- leucine rich repeat and fibronectin type III domain containing 3
View Full Product Details