Product Details

SNP ID
rs141797988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54279970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGATTTTTTCTCCCTATATAGA[C/T]GGTACTCCTGGGCTTCAAGGCTCCC
Phenotype
MIM: 604815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LILRB2 PubMed Links

Gene Details

Gene
LILRB2
Gene Name
leukocyte immunoglobulin like receptor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080978.3 Intron NP_001074447.2
NM_001278403.2 Intron NP_001265332.2
NM_001278404.2 Intron NP_001265333.2
NM_001278405.2 Intron NP_001265334.2
NM_001278406.2 Intron NP_001265335.2
NM_005874.4 Intron NP_005865.3
Gene
MIR4752
Gene Name
microRNA 4752
There are no transcripts associated with this gene.

View Full Product Details