Product Details

SNP ID
rs144123905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6752635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCTGCGACAGGACGCCGAGGAC[A/G]CGCTGGAACTTCTCAAAGCGTTCAG
Phenotype
MIM: 604721 MIM: 604504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH2D3A PubMed Links

Gene Details

Gene
SH2D3A
Gene Name
SH2 domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005490.2 1560 Silent Mutation CGC,CGT R563R NP_005481.2
XM_005259467.1 1560 Silent Mutation CGC,CGT R592R XP_005259524.1
XM_005259472.2 1560 Silent Mutation CGC,CGT R470R XP_005259529.1
XM_006722602.1 1560 Silent Mutation CGC,CGT R472R XP_006722665.1
XM_011527608.2 1560 Silent Mutation CGC,CGT R497R XP_011525910.1
XM_011527609.2 1560 Silent Mutation CGC,CGT R497R XP_011525911.1
XM_017026123.1 1560 Intron XP_016881612.1
XM_017026124.1 1560 Intron XP_016881613.1
Gene
TRIP10
Gene Name
thyroid hormone receptor interactor 10
There are no transcripts associated with this gene.

View Full Product Details