Product Details

SNP ID
rs144505058
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.19:11947568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAGTACCAAAACCCCAGAAGAA[A/G]CTTCAGGTAATTTGCATTTCCAAGA
Phenotype
MIM: 194543
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF69 PubMed Links

Gene Details

Gene
ZNF69
Gene Name
zinc finger protein 69
There are no transcripts associated with this gene.

Gene
ZNF700
Gene Name
zinc finger protein 700
There are no transcripts associated with this gene.

View Full Product Details