Product Details

SNP ID
rs139771818
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:171784129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTCCCGGGAGGGGTGGATGGT[A/G]AGGGTGCACCAAGTGGCCAACAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL13 PubMed Links

Gene Details

Gene
METTL13
Gene Name
methyltransferase like 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007239.1 557 Intron NP_001007240.1
NM_014955.2 557 Silent Mutation GTA,GTG V95V NP_055770.1
NM_015935.4 557 Silent Mutation GTA,GTG V181V NP_057019.3
XM_017001455.1 557 Silent Mutation GTA,GTG V95V XP_016856944.1
XM_017001456.1 557 Silent Mutation GTA,GTG V95V XP_016856945.1
XM_017001457.1 557 Silent Mutation GTA,GTG V95V XP_016856946.1
XM_017001458.1 557 Silent Mutation GTA,GTG V181V XP_016856947.1
XM_017001459.1 557 Silent Mutation GTA,GTG V95V XP_016856948.1

View Full Product Details