Product Details

SNP ID
rs144116274
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:171782087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAACCTACCTGGAACTGTGCGG[C/G]GTGCTACATAAATATATCAAGCCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
METTL13 PubMed Links

Gene Details

Gene
METTL13
Gene Name
methyltransferase like 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007239.1 467 Silent Mutation GGC,GGG G40G NP_001007240.1
NM_014955.2 467 Intron NP_055770.1
NM_015935.4 467 Silent Mutation GGC,GGG G40G NP_057019.3
XM_017001455.1 467 Intron XP_016856944.1
XM_017001456.1 467 Intron XP_016856945.1
XM_017001457.1 467 Intron XP_016856946.1
XM_017001458.1 467 Silent Mutation GGC,GGG G40G XP_016856947.1
XM_017001459.1 467 Intron XP_016856948.1

View Full Product Details