Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267609.1 | 2074 | Missense Mutation | NP_001254538.1 | ||
NM_001267610.1 | 2074 | Missense Mutation | NP_001254539.1 | ||
NM_001267611.1 | 2074 | Missense Mutation | NP_001254540.1 | ||
NM_001267612.1 | 2074 | Missense Mutation | NP_001254541.1 | ||
NM_001267613.1 | 2074 | Missense Mutation | NP_001254542.1 | ||
NM_001267614.1 | 2074 | Missense Mutation | NP_001254543.1 | ||
NM_153832.2 | 2074 | Missense Mutation | NP_722561.1 | ||
XM_005245055.2 | 2074 | Nonsense Mutation | XP_005245112.1 | ||
XM_005245056.2 | 2074 | Nonsense Mutation | XP_005245113.1 | ||
XM_005245057.4 | 2074 | Nonsense Mutation | XP_005245114.1 | ||
XM_006711251.1 | 2074 | Nonsense Mutation | XP_006711314.1 | ||
XM_006711253.2 | 2074 | Nonsense Mutation | XP_006711316.2 | ||
XM_011509370.1 | 2074 | Nonsense Mutation | XP_011507672.1 | ||
XM_011509371.2 | 2074 | Nonsense Mutation | XP_011507673.1 | ||
XM_011509372.1 | 2074 | Nonsense Mutation | XP_011507674.1 | ||
XM_011509373.2 | 2074 | Nonsense Mutation | XP_011507675.1 | ||
XM_011509375.2 | 2074 | Nonsense Mutation | XP_011507677.1 | ||
XM_011509376.2 | 2074 | Nonsense Mutation | XP_011507678.1 | ||
XM_011509377.1 | 2074 | Nonsense Mutation | XP_011507679.1 | ||
XM_011509378.1 | 2074 | Nonsense Mutation | XP_011507680.1 | ||
XM_017000858.1 | 2074 | Nonsense Mutation | XP_016856347.1 | ||
XM_017000859.1 | 2074 | Nonsense Mutation | XP_016856348.1 | ||
XM_017000860.1 | 2074 | Nonsense Mutation | XP_016856349.1 | ||
XM_017000861.1 | 2074 | Nonsense Mutation | XP_016856350.1 | ||
XM_017000862.1 | 2074 | Nonsense Mutation | XP_016856351.1 |