Product Details
- SNP ID
-
rs146918615
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38080939 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGCGCCCACAGCCGCCATGAGCA[C/T]CTCGAACTGCAGCGCGCCCACCATG
- Phenotype
-
MIM: 605926
MIM: 610409
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BAIAP2L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138209611] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAIAP2L2
- Gene Name
- BAI1 associated protein 2 like 2
There are no transcripts associated with this gene.
- Gene
- PICK1
- Gene Name
- protein interacting with PRKCA 1
There are no transcripts associated with this gene.
- Gene
- SLC16A8
- Gene Name
- solute carrier family 16 member 8
View Full Product Details