Product Details

SNP ID
rs150102347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140535813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTATGAAGGAAAAGAATATTAA[A/C]CCTGAGAAGGATGCCAGGTGTGCGT
Phenotype
MIM: 610196
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELMOD2 PubMed Links

Gene Details

Gene
ELMOD2
Gene Name
ELMO domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153702.3 741 Missense Mutation AAA,AAC K84N NP_714913.1
XM_005262885.3 741 Missense Mutation AAA,AAC K96N XP_005262942.1
XM_005262886.2 741 Missense Mutation AAA,AAC K84N XP_005262943.1
XM_011531818.2 741 Missense Mutation AAA,AAC K96N XP_011530120.1
XM_011531819.2 741 Missense Mutation AAA,AAC K84N XP_011530121.1

View Full Product Details