Product Details

SNP ID
rs137949520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172769789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAAGGGCAGGATTTCCACCGG[A/G]CCACCCTGAAATCCAGAAATATCCA
Phenotype
MIM: 600714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP1 PubMed Links

Gene Details

Gene
DUSP1
Gene Name
dual specificity phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004417.3 767 Silent Mutation GGC,GGT G173G NP_004408.1
Gene
LOC101928093
Gene Name
uncharacterized LOC101928093
There are no transcripts associated with this gene.

View Full Product Details