Product Details

SNP ID
rs149605403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:91374274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGCTCCAGGAATATCCACATA[C/T]ACCTAAACATTGCAAAGAAATATTA
Phenotype
MIM: 612464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARRDC3 PubMed Links

Gene Details

Gene
ARRDC3
Gene Name
arrestin domain containing 3
There are no transcripts associated with this gene.

Gene
ARRDC3-AS1
Gene Name
ARRDC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details