Product Details
- SNP ID
-
rs143506536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130346522 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGAGGCGGGACGCGCCCTGGG[C/T]CATCCCCCGTGGACAGGCGGACACT
- Phenotype
-
MIM: 609561
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPA5
PubMed Links
Gene Details
- Gene
- CPA5
- Gene Name
- carboxypeptidase A5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127441.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
NP_001120913.1 |
NM_001127442.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
NP_001120914.1 |
NM_001318223.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
NP_001305152.1 |
NM_080385.4 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
NP_525124.3 |
XM_005250710.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_005250767.1 |
XM_005250712.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_005250769.1 |
XM_011516698.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_011515000.1 |
XM_011516701.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_011515003.1 |
XM_011516702.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_011515004.1 |
XM_011516703.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_011515005.1 |
XM_011516704.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_011515006.1 |
XM_011516705.1 |
1125 |
Missense Mutation |
CCA,TCA |
P13S |
XP_011515007.1 |
- Gene
- LOC101928632
- Gene Name
- uncharacterized LOC101928632
There are no transcripts associated with this gene.
View Full Product Details