Product Details

SNP ID
rs145871579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35100053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCACTAACTCATCTTGACTCG[A/G]TCAAGTTCCTCATCAAGACTTGCAT
Phenotype
MIM: 614730 MIM: 608292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM214B PubMed Links

Gene Details

Gene
FAM214B
Gene Name
family with sequence similarity 214 member B
There are no transcripts associated with this gene.

Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
There are no transcripts associated with this gene.

Gene
STOML2
Gene Name
stomatin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287031.1 1043 Silent Mutation GAC,GAT D306D NP_001273960.1
NM_001287032.1 1043 Silent Mutation GAC,GAT D300D NP_001273961.1
NM_001287033.1 1043 Silent Mutation GAC,GAT D305D NP_001273962.1
NM_013442.2 1043 Silent Mutation GAC,GAT D351D NP_038470.1

View Full Product Details