Product Details

SNP ID
rs199475795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:279481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGGCTGGAGCGCGCGGACGCC[A/G]TGGACCTCGCGGAGCAGCTGGCCCA
Phenotype
MIM: 609650 MIM: 617032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRP6 PubMed Links

Gene Details

Gene
NLRP6
Gene Name
NLR family pyrin domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276700.1 389 Missense Mutation ATG,GTG M62V NP_001263629.1
NM_138329.2 389 Missense Mutation ATG,GTG M62V NP_612202.2
XM_011519931.1 389 Missense Mutation ATG,GTG M62V XP_011518233.1
XM_017017252.1 389 Missense Mutation ATG,GTG M62V XP_016872741.1
XM_017017253.1 389 Intron XP_016872742.1
Gene
PGGHG
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
There are no transcripts associated with this gene.

View Full Product Details