Product Details

SNP ID
rs200902043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52544611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTGTAATCGTTGGCACTGCCC[C/T]GGCTCCTGCCCTCCCCGCCTCTCAC
Phenotype
MIM: 608245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT71 PubMed Links

Gene Details

Gene
KRT71
Gene Name
keratin 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033448.2 1563 Missense Mutation CAG,CGG Q498R NP_258259.1
XM_017018749.1 1563 Missense Mutation CAG,CGG Q416R XP_016874238.1

View Full Product Details