Product Details

SNP ID
rs202112087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52544636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCCTGCCCTCCCCGCCTCTCA[C/T]GCTGCACACTCCAGAGATGCAGTTG
Phenotype
MIM: 608245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT71 PubMed Links

Gene Details

Gene
KRT71
Gene Name
keratin 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033448.2 1538 Missense Mutation ATG,GTG M490V NP_258259.1
XM_017018749.1 1538 Missense Mutation ATG,GTG M408V XP_016874238.1

View Full Product Details