Product Details

SNP ID
rs200464088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59207967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTATTCCTCAGTATTCCTTGT[A/G]TCCTGGGAGAGAACGGTATTACCAA
Phenotype
MIM: 601479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDHAL6B PubMed Links

Gene Details

Gene
LDHAL6B
Gene Name
lactate dehydrogenase A like 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033195.2 1152 Missense Mutation ATC,GTC I343V NP_149972.1
Gene
MYO1E
Gene Name
myosin IE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004998.3 1152 Intron NP_004989.2

View Full Product Details