Product Details

SNP ID
rs201654205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2853884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACCAGACAAGCATCCCGCTCC[C/G]CCTCCAAGTAGCCGGCACACAGCAT
Phenotype
MIM: 609343
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS22 PubMed Links

Gene Details

Gene
PRSS22
Gene Name
protease, serine 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022119.3 764 Missense Mutation GCG,GGG A233G NP_071402.1
XM_005255473.3 764 Missense Mutation GCG,GGG A233G XP_005255530.1

View Full Product Details